4-(Trifluoromethoxy)toluene 98%

Supplier: Ambeed
Danger

A760913-25G A760913-5G
77146-310EA 57.29 USD
77146-310 77146-312
4-(Trifluoromethoxy)toluene 98%
4-(Trifluoromethoxy)toluene

Precaución: for Research Use Only

Formula: C₈H₇F₃O
MDL Number: MFCD00042413
CAS Number: 706-27-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR