1-(2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N,N-dimethylmethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A760910-100MG A760910-1G A760910-250MG
77146-304EA 36.08 USD
77146-304 77146-306 77146-308
1-(2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N,N-dimethylmethanamine hydrochloride 97%
1-(2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N,N-dimethylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₄BClFNO₂
MW: 315.61 g/mol
MDL Number: MFCD20441914
CAS Number: 2096332-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR