(R)-N-(1-Benzylpyrrolidin-3-yl)acetamide 98+%

Supplier: Ambeed
Warning

A758324-100MG A758324-1G A758324-250MG
77146-284EA 41.38 USD
77146-284 77146-286 77146-288
(R)-N-(1-Benzylpyrrolidin-3-yl)acetamide 98+%
(R)-N-(1-Benzylpyrrolidin-3-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O
MW: 218.29 g/mol
MDL Number: MFCD04974061
CAS Number: 114636-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR