(R)-4-(Hydroxymethyl)oxazolidin-2-one 98%
Supplier: Ambeed
|
Warning
|
A757628-100MG
A757628-1G
A757628-250MG
77146-278EA
67.9
USD
77146-278
77146-280
77146-282
(R)-4-(Hydroxymethyl)oxazolidin-2-one 98%
(R)-4-(Hydroxymethyl)oxazolidin-2-one
Precaución: for Research Use Only
Formula:
C₄H₇NO₃ MW: 117.10 g/mol |
MDL Number:
MFCD11847629 CAS Number: 132682-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...