(-)-Diacetyl-D-tartaric anhydride 98%

Supplier: Ambeed
Warning

Synonyms: (3S,4S)-2,5-Dioxotetrahydrofuran-3,4-diyl diacetate

A757259-25G A757259-5G
77146-274EA 180.24 USD
77146-274 77146-276
(-)-Diacetyl-D-tartaric anhydride 98%
(-)-Diacetyl-D-tartaric anhydride

Precaución: for Research Use Only

Formula: C₈H₈O₇
MDL Number: MFCD02093461
CAS Number: 70728-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR