5,7-Dichloro-3-iodopyrazolo[1,5-a]pyrimidine 98%

Supplier: Ambeed
Warning

A754259-100MG A754259-250MG
77146-258EA 99.73 USD
77146-258 77146-260
5,7-Dichloro-3-iodopyrazolo[1,5-a]pyrimidine 98%
5,7-Dichloro-3-iodopyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂IN₃
MW: 313.91 g/mol
MDL Number: MFCD18073831
CAS Number: 754211-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR