2-Fluoro-5-formylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-formylbenzeneboronic acid

A750784-100G A750784-1G A750784-25G A750784-5G
77146-246EA 545.31 USD
77146-246 77146-248 77146-250 77146-252
2-Fluoro-5-formylphenylboronic acid 98%
2-Fluoro-5-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₃
MW: 167.93 g/mol
MDL Number: MFCD03095100
CAS Number: 352534-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR