(2,4-Dichloro-6-hydroxyphenyl)boronic acid 97%
Supplier: Ambeed
|
Warning
|
A750278-100MG
A750278-250MG
77146-242EA
79.31
USD
77146-242
77146-244
(2,4-Dichloro-6-hydroxyphenyl)boronic acid 97%
(2,4-Dichloro-6-hydroxyphenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₆H₅BCl₂O₃ |
MDL Number:
MFCD12031243 CAS Number: 1028332-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...