6-Bromo-7-fluoroindoline-2,3-dione 97%

Supplier: Ambeed
Warning

A749837-100MG A749837-10G A749837-250MG A749837-5G
77146-234EA 56.23 USD
77146-234 77146-236 77146-238 77146-240
6-Bromo-7-fluoroindoline-2,3-dione 97%
6-Bromo-7-fluoroindoline-2,3-dione

Precaución: for Research Use Only

Formula: C₈H₃BrFNO₂
MW: 244.01 g/mol
MDL Number: MFCD27943837
CAS Number: 1336963-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR