3-Carboxy-4-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Carboxy-4-fluorophenylboronic Acid, 3-Carboxy-4-fluorobenzeneboronic acid

A749251-1G A749251-25G A749251-5G
77146-222EA 11.67 USD
77146-222 77146-224 77146-226
3-Carboxy-4-fluorophenylboronic acid 98%
3-Carboxy-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₄
MW: 183.93 g/mol
MDL Number: MFCD03095129
CAS Number: 872460-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR