4-Ethoxy-3-fluorophenol 97%

Supplier: Ambeed
Warning

A748949-100MG A748949-1G A748949-250MG
77146-216EA 32.96 USD
77146-216 77146-218 77146-220
4-Ethoxy-3-fluorophenol 97%
4-Ethoxy-3-fluorophenol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MDL Number: MFCD21984328
CAS Number: 98121-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR