2-(3-Chloro-4-(cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A748018-100MG A748018-1G A748018-250MG A748018-5G
77146-208EA 37.14 USD
77146-208 77146-210 77146-212 77146-214
2-(3-Chloro-4-(cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3-Chloro-4-(cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₂BClO₃
MW: 308.61 g/mol
MDL Number: MFCD25953905
CAS Number: 1860005-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR