(4-Fluorobenzyl)triphenylphosphonium bromide 97%
Supplier: Ambeed
|
Warning
|
A746071-5G
A746071-25G
A746071-1G
77146-202EA
43.25
USD
77146-202
77595-176
77747-472
(4-Fluorobenzyl)triphenylphosphonium bromide 97%
(4-Fluorobenzyl)triphenylphosphonium bromide
Precaución: for Research Use Only
Formula:
C₂₅H₂₁BrFP |
MDL Number:
MFCD00032509 CAS Number: 51044-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...