1-Bromo-5-methoxy-4-methyl-2-nitrobenzene 98%

Supplier: Ambeed
Warning

A745869-1G A745869-250MG A745869-5G
77146-196EA 93.36 USD
77146-196 77146-198 77146-200
1-Bromo-5-methoxy-4-methyl-2-nitrobenzene 98%
1-Bromo-5-methoxy-4-methyl-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.05 g/mol
MDL Number: MFCD28291981
CAS Number: 1807224-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR