Boc-DL-Cha-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-((tert-Butoxycarbonyl)amino)-3-cyclohexylpropanoic acid
A745795-1G
A745795-5G
77146-192EA
18.04
USD
77146-192
77146-194
Boc-DL-Cha-OH 98%
Boc-DL-Cha-OH
Precaución: for Research Use Only
Formula:
C₁₄H₂₅NO₄ MW: 271.353 g/mol |
MDL Number:
MFCD03095514 CAS Number: 144186-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...