5-Chloro-2-methoxyphenylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Carboxymethyl)-4-chloroanisole, (5-Chloro-2-methoxyphenyl)acetic Acid

A744862-1G A744862-25G A744862-5G
77146-184EA 25.47 USD
77146-184 77146-186 77146-188
5-Chloro-2-methoxyphenylacetic acid 98%
5-Chloro-2-methoxyphenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.619 g/mol
MDL Number: MFCD02664695
CAS Number: 7569-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR