(4R,6S)-6-Methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4R,6S)-6-Methyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol-7,7-dioxide
A743824-100G
A743824-10G
A743824-1G
A743824-25G
A743824-5G
77146-168EA
623.81
USD
77146-168
77146-170
77146-172
77146-174
77146-176
(4R,6S)-6-Methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol 98%
(4R,6S)-6-Methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol
Precaución: for Research Use Only
Formula:
C₈H₁₀O₃S₂ MW: 218.29 g/mol |
MDL Number:
MFCD11035925 CAS Number: 147128-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...