(4-Cyano-3-isobutoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A743637-1G A743637-250MG A743637-5G
77146-162EA 156.57 USD
77146-162 77146-164 77146-166
(4-Cyano-3-isobutoxyphenyl)boronic acid 97%
(4-Cyano-3-isobutoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄BNO₃
MDL Number: MFCD19684100
CAS Number: 2096335-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR