tert-Butyl 1H-indol-4-ylcarbamate 95%

Supplier: Ambeed
Warning

A742596-100MG A742596-1G A742596-250MG
77146-156EA 58.35 USD
77146-156 77146-158 77146-160
tert-Butyl 1H-indol-4-ylcarbamate 95%
tert-Butyl 1H-indol-4-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
MDL Number: MFCD04973303
CAS Number: 819850-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR