(S)-(+)-2-(Methoxymethyl)pyrrolidine 97%

Supplier: Ambeed
Danger

Synonyms: (S)-2-(Methoxymethyl)pyrrolidine

A742383-1G A742383-5G A742383-250MG
77146-152EA 33.95 USD
77146-152 77146-154 77747-465
(S)-(+)-2-(Methoxymethyl)pyrrolidine 97%
(S)-(+)-2-(Methoxymethyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD00010408
CAS Number: 63126-47-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR