(S)-(-)-2-Methyl-CBS-oxazaborolidine monohydrate 97% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: (S)-1-Methyl-3,3-diphenylhexahydropyrrolo[1,2-c][1,3,2]oxazaborole

A742072-100G A742072-10G A742072-1G A742072-25G A742072-500G A742072-5G
77146-140EA 167.63 USD
77146-140 77146-142 77146-144 77146-146 77146-148 77146-150
(S)-(-)-2-Methyl-CBS-oxazaborolidine monohydrate 97% (by HPLC)
(S)-(-)-2-Methyl-CBS-oxazaborolidine monohydrate

Precaución: for Research Use Only

Formula: C₁₈H₂₀BNO·1H₂O
MW: 295.19 g/mol (277.17 anhy)
MDL Number: MFCD00078439
CAS Number: 112022-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR