Methyl 2,3,4-trifluoro-5-nitrobenzoate 98%

Supplier: Ambeed
Warning

A741334-1G A741334-250MG A741334-5G
77146-128EA 33.98 USD
77146-128 77146-130 77146-132
Methyl 2,3,4-trifluoro-5-nitrobenzoate 98%
Methyl 2,3,4-trifluoro-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₄
MDL Number: MFCD13248633
CAS Number: 918321-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR