Diethyl 2-fluoro-2-methylmalonate 95%

Supplier: Ambeed
Danger

A739666-10G A739666-1G A739666-250MG A739666-25G A739666-5G
77146-118EA 698.08 USD
77146-118 77146-120 77146-122 77146-124 77146-126
Diethyl 2-fluoro-2-methylmalonate 95%
Diethyl 2-fluoro-2-methylmalonate

Precaución: for Research Use Only

Formula: C₈H₁₃FO₄
MW: 192.19 g/mol
MDL Number: MFCD03844748
CAS Number: 16519-02-1
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR