5-Chloro-2,3-difluorobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A739614-1G
A739614-250MG
A739614-5G
77146-114EA
117.41
USD
77146-114
77146-116
77686-747
5-Chloro-2,3-difluorobenzaldehyde 97%
5-Chloro-2,3-difluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₃ClF₂O |
MDL Number:
MFCD20924197 CAS Number: 1783512-25-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...