Methyl 2,6-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97%

Supplier: Ambeed
Warning

A737652-1G A737652-250MG A737652-5G
77146-098EA 207.94 USD
77146-098 77146-100 77146-102
Methyl 2,6-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97%
Methyl 2,6-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₆H₂₃BO₄
MW: 290.16 g/mol
MDL Number: MFCD18311856
CAS Number: 2096333-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR