2,5-Dioxopyrrolidin-1-yl 2,2-dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oate 99%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG4-NHS ester

A737346-250MG A737346-1G A737346-100MG A737346-5G
77146-094EA 45.32 USD
77146-094 77618-032 77686-744 77686-745
2,5-Dioxopyrrolidin-1-yl 2,2-dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oate 99%
2,5-Dioxopyrrolidin-1-yl 2,2-dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oate

Precaución: for Research Use Only

Formula: C₂₀H₃₄N₂O₁₀
MDL Number: MFCD20228887
CAS Number: 859230-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR