2-Iodo-5-nitrobenzonitrile 97%

Supplier: Ambeed
Danger

A735782-1G A735782-5G
77146-090EA 30.77 USD
77146-090 77146-092
2-Iodo-5-nitrobenzonitrile 97%
2-Iodo-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃IN₂O₂
MW: 274.01 g/mol
MDL Number: MFCD11521302
CAS Number: 101421-15-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR