N,N'-Bis[4-di(m-tolyl)aminophenyl]-N,N'-diphenylbenzidine 97%

Supplier: Ambeed
Warning

Synonyms: N1,N1'-([1,1'-Biphenyl]-4,4'-diyl)bis(N1-phenyl-N4,N4-di-m-tolylbenzene-1,4-diamine), DNTPD

A732474-1G A732474-250MG
77146-080EA 46.35 USD
77146-080 77146-082
N,N'-Bis[4-di(m-tolyl)aminophenyl]-N,N'-diphenylbenzidine 97%
N,N'-Bis[4-di(m-tolyl)aminophenyl]-N,N'-diphenylbenzidine

Precaución: for Research Use Only

Formula: C₆₄H₅₄N₄
MW: 879.16 g/mol
MDL Number: MFCD22372686
CAS Number: 199121-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR