Amino-PEG5-Amine 98%

Supplier: Ambeed
Warning

A729766-100MG A729766-1G A729766-250MG A729766-5G
77146-054EA 25.75 USD
77146-054 77146-056 77146-058 77146-060
Amino-PEG5-Amine 98%
Amino-PEG5-Amine

Precaución: for Research Use Only

Formula: C₁₂H₂₈N₂O₅
MDL Number: MFCD07369626
CAS Number: 72236-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR