2-Chloro-4-formylbenzoic acid 97%

Supplier: Ambeed
Warning

A728538-1G A728538-250MG A728538-100MG
77146-042EA 204.76 USD
77146-042 77146-044 77747-439
2-Chloro-4-formylbenzoic acid 97%
2-Chloro-4-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅ClO₃
MW: 184.57 g/mol
MDL Number: MFCD11110998
CAS Number: 1289063-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR