3-(Benzyloxy)-4-oxo-4H-pyran-2-carbaldehyde 97%

Supplier: Ambeed
Warning

A727814-100MG A727814-1G A727814-250MG A727814-5G
77146-028EA 77.45 USD
77146-028 77146-030 77146-032 77146-034
3-(Benzyloxy)-4-oxo-4H-pyran-2-carbaldehyde 97%
3-(Benzyloxy)-4-oxo-4H-pyran-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₄
MW: 230.22 g/mol
MDL Number: MFCD29921839
CAS Number: 500371-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR