4,6-Dichloro-3-iodo-1H-pyrazolo[4,3-c]pyridine 97%

Supplier: Ambeed
Warning

A725041-100MG A725041-250MG
77146-004EA 23.34 USD
77146-004 77146-006
4,6-Dichloro-3-iodo-1H-pyrazolo[4,3-c]pyridine 97%
4,6-Dichloro-3-iodo-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂IN₃
MW: 313.91 g/mol
MDL Number: MFCD29921795
CAS Number: 1658467-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR