(R)-1-Boc-3-Propylpiperazine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (R)-3-propylpiperazine-1-carboxylate

A724204-1G A724204-250MG A724204-5G
77145-992EA 89.12 USD
77145-992 77145-994 77145-996
(R)-1-Boc-3-Propylpiperazine 97%
(R)-1-Boc-3-propylpiperazine

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
MW: 228.3312 g/mol
MDL Number: MFCD07772107
CAS Number: 928025-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR