Diethyl (3-chlorobenzyl)phosphonate 98%

Supplier: Ambeed
Warning

A722964-1G A722964-25G A722964-5G
77145-982EA 19.1 USD
77145-982 77145-984 77617-960
Diethyl (3-chlorobenzyl)phosphonate 98%
Diethyl (3-chlorobenzyl)phosphonate

Precaución: for Research Use Only

Formula: C₁₁H₁₆ClO₃P
MDL Number: MFCD01861282
CAS Number: 78055-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR