1-Methylsulfonyl-1H-benzotriazole 97%

Supplier: Ambeed
Danger

Synonyms: 1-(Methylsulphonyl)-1H-benzo[d][1,2,3]triazole

A721268-250MG A721268-25G A721268-5G
77145-970EA 12.74 USD
77145-970 77145-972 77145-974
1-Methylsulfonyl-1H-benzotriazole 97%
1-Methylsulfonyl-1H-benzotriazole

Precaución: for Research Use Only

Formula: C₇H₇N₃O₂S
MW: 197.21 g/mol
MDL Number: MFCD00595638
CAS Number: 37073-15-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR