(2S,4R)-1-[(tert-Butoxy)carbonyl]-4-methylpyrrolidine-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,4R)-1-(tert-Butoxycarbonyl)-4-methylpyrrolidine-2-carboxylic acid, (2S,4R)-4-Methylpyrrolidine-2-carboxylic acid, N-BOC protected
A719790-100MG
A719790-1G
A719790-250MG
77145-964EA
144.29
USD
77145-964
77145-966
77145-968
(2S,4R)-1-[(tert-Butoxy)carbonyl]-4-methylpyrrolidine-2-carboxylic acid 97%
(2S,4R)-1-[(tert-Butoxy)carbonyl]-4-methylpyrrolidine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol |
MDL Number:
MFCD13185988 CAS Number: 364750-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...