CW069 99%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(2-(Benzylamino)-3-phenylpropanamido)-5-iodobenzoic acid

A719211-10MG A719211-1MG A719211-2MG A719211-5MG
77145-956EA 154.9 USD
77145-956 77145-958 77145-960 77145-962
CW069 99%
CW069

Precaución: for Research Use Only

Formula: C₂₃H₂₁IN₂O₃
MW: 500.33 g/mol
MDL Number: MFCD28009372
CAS Number: 1594094-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR