(1S,2S)-(-)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine 97%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-N,N'-Dimethyl-1,2-diphenylethane-1,2-diamine

A716988-100MG A716988-1G A716988-250MG A716988-25G A716988-5G
77145-946EA 22.28 USD
77145-946 77145-948 77145-950 77145-952 77145-954
(1S,2S)-(-)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine 97%
(1S,2S)-(-)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
MDL Number: MFCD00216956
CAS Number: 70749-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR