Tris(triphenylphosphine)copper(I) bromide 98%

Supplier: Ambeed
Warning

Synonyms: Bromotris(triphenylphosphine)copper(I)

A716803-100G A716803-1G A716803-25G A716803-5G
77145-940EA 448.05 USD
77145-940 77145-942 77145-944 77686-705
Tris(triphenylphosphine)copper(I) bromide 98%
Tris(triphenylphosphine)copper(I) bromide

Precaución: for Research Use Only

Formula: C₅₄H₄₅BrCuP₃
MW: 930.32 g/mol
MDL Number: MFCD04118115
CAS Number: 15709-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR