AMG-9810 98%

Supplier: Ambeed
Warning

A715034-100MG A715034-10MG A715034-50MG A715034-5MG A715034-1MG
77145-928EA 119.47 USD
77145-928 77145-930 77145-932 77145-934 77617-940
AMG-9810 98%
AMG-9810

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₃
MDL Number: MFCD00955103
CAS Number: 545395-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR