6-Chloro-1-methyl-1H-pyrazolo[4,3-c]pyridine 97%
Supplier: Ambeed
|
Danger
|
A714261-100MG
A714261-1G
77145-926EA
74.27
USD
77145-926
77747-417
6-Chloro-1-methyl-1H-pyrazolo[4,3-c]pyridine 97%
6-Chloro-1-methyl-1H-pyrazolo[4,3-c]pyridine
Precaución: for Research Use Only
Formula:
C₇H₆ClN₃ MW: 167.596 g/mol |
MDL Number:
MFCD28608322 CAS Number: 1558302-68-3 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...