(S,S)-(+)-2,4-Pentanediol 98%

Supplier: Ambeed
Warning

A713940-100MG A713940-10G A713940-1G A713940-250MG A713940-25G A713940-5G
77145-914EA 17.51 USD
77145-914 77145-916 77145-918 77145-920 77145-922 77145-924
(S,S)-(+)-2,4-Pentanediol 98%
(S,S)-(+)-2,4-Pentanediol

Precaución: for Research Use Only

Formula: C₅H₁₂O₂
MDL Number: MFCD00064276
CAS Number: 72345-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR