6-Chloro-3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-8-amine 97%

Supplier: Ambeed
Warning

A713729-100MG A713729-1G A713729-250MG A713729-5G
77145-906EA 19.57 USD
77145-906 77145-908 77145-910 77145-912
6-Chloro-3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-8-amine 97%
6-Chloro-3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-8-amine

Precaución: for Research Use Only

Formula: C₆H₆ClN₅
MDL Number: MFCD20707049
CAS Number: 53085-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR