(2,4-Pentanedionato)bis(2-phenylpyridine)iridium(III) ≥98%

Supplier: Ambeed
Warning

A711748-100MG A711748-250MG A711748-1G
77145-888EA 48.41 USD
77145-888 77145-890 77686-698
(2,4-Pentanedionato)bis(2-phenylpyridine)iridium(III) ≥98%
(2,4-Pentanedionato)bis(2-phenylpyridine)iridium(III)

Precaución: for Research Use Only

Formula: C₂₇H₂₃IrN₂O₂
MDL Number: MFCD12022553
CAS Number: 337526-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR