(4-Bromo-3-methyl-1H-pyrazol-5-yl)methanol 97%

Supplier: Ambeed
Warning

A711000-1G A711000-250MG
77145-884EA 110.21 USD
77145-884 77145-886
(4-Bromo-3-methyl-1H-pyrazol-5-yl)methanol 97%
(4-Bromo-3-methyl-1H-pyrazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₇BrN₂O
MDL Number: MFCD28405190
CAS Number: 2828447-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR