4-Boronobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 4-Sulphamoylbenzeneboronic acid, (4-Aminosulphonylphenyl)boronic acid

A706846-100MG A706846-10G A706846-1G A706846-250MG A706846-25G A706846-5G
77145-842EA 15.92 USD
77145-842 77145-844 77145-846 77145-848 77145-850 77145-852
4-Boronobenzenesulfonamide 98%
4-Boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₈BNO₄S
MW: 201.00 g/mol
MDL Number: MFCD06659839
CAS Number: 613660-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR