(4-Bromophenyl)methanesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Bromobenzylsulphonyl chloride

A706084-1G A706084-250MG A706084-5G
77145-830EA 40.32 USD
77145-830 77145-832 77145-834
(4-Bromophenyl)methanesulfonyl chloride 98%
(4-Bromophenyl)methanesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆BrClO₂S
MW: 269.5433 g/mol
MDL Number: MFCD04973454
CAS Number: 53531-69-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR