2-Bromo-3-fluoro-6-hydroxybenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A705702-100MG
A705702-250MG
A705702-50MG
77145-820EA
51.51
USD
77145-820
77145-822
77145-824
2-Bromo-3-fluoro-6-hydroxybenzaldehyde 98%
2-Bromo-3-fluoro-6-hydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄BrFO₂ |
MDL Number:
MFCD23705549 CAS Number: 1427382-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...