3-Methoxybutyl thioglycolate 97% (by GC)

Supplier: Ambeed
Danger

Synonyms: 3-Methoxybutyl 2-mercaptoacetate, 3-Methoxybutyl Mercaptoacetate, Mercaptoacetic Acid 3-Methoxybutyl Ester, Thioglycolic Acid 3-Methoxybutyl Ester

A704431-25G A704431-5G
77145-816EA 44.28 USD
77145-816 77145-818
3-Methoxybutyl thioglycolate 97% (by GC)
3-Methoxybutyl thioglycolate

Precaución: for Research Use Only

Formula: C₇H₁₄O₃S
MW: 178.25 g/mol
MDL Number: MFCD04038084
CAS Number: 27431-39-6
UN: 3334
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR