1-(2-Bromo-4-chloropyridin-3-yl)ethanone 97%

Supplier: Ambeed
Warning

A704282-1G A704282-250MG
77145-812EA 331.01 USD
77145-812 77145-814
1-(2-Bromo-4-chloropyridin-3-yl)ethanone 97%
1-(2-Bromo-4-chloropyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅BrClNO
MW: 234.47 g/mol
MDL Number: MFCD29058979
CAS Number: 1956319-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR